
Adenine hydrochloride
Catalog # MOLE85430679-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:171.59 g/mol
EINECS:220-867-0
:GHS07
CAS Number:2922-28-3
:6055-72-7
Formula:C₅H₅N₅·HCl
MDL Number:MFCD00150864