
Guanidinium thiocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
Melting Pt:118 °C
Density:1.29 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:209-812-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together