

Guanidinium thiocyanate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
Melting Pt:118 °C
Density:1.29 g/cm³ (20 °C)
EINECS:209-812-1
UN:1759
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1420 g/l (20 °C)
- Melting Point:118 °C
- Molar Mass:118.16 g/mol
- Bulk Density:630 kg/m3
- Density:1.29 g/cm3 (20 °C)
- pH:4.8 - 6.0 (1420 g/l, H2O, 20 °C) (saturated solution)
- Hazard Class:8
- Hazard Class Description:Corrosive Material