Order Entry
Germany
ContactUsLinkComponent
Guanidinium carbonate for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Guanidinium carbonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Guanidinium carbonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    500 g/l (30 °C)
  • Melting Point:
    197 °C
  • Molar Mass:
    180.17 g/mol
  • Bulk Density:
    500 - 700 kg/m3
  • Density:
    1.25 g/cm3 (20 °C)
  • pH:
    11.5 (110 g/l, H2O, 20 °C)
  • Ignition Point:
    530 °C
  • Hill Formula:
    C3H12N6O3
  • Chemical Formula:
    (CH5N3)2 * H2CO3

Specification Test Results

Related Information

Certificates
Search by Lot Number