Spermidine trihydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:334-50-9
:
Formula:C₇H₂₂Cl₃N₃
MDL Number:MFCD00012918
:254.63 g/mol
Merck Index:12,08893
Melting Pt:256…258 °C
Storage Temperature:Freezer
EINECS:206-379-0