Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spermidine trihydrochloride
Spermidine trihydrochloride
Spermidine trihydrochloride
Spermidine trihydrochloride
Supplier:
Serva
Certificates
Expand image
Spermidine trihydrochloride
Supplier:
Serva
About this item
CAS Number:
334-50-9
:
Formula:
C₇H₂₂Cl₃N₃
MDL Number:
MFCD00012918
:
254.63 g/mol
Merck Index:
12,08893
Melting Pt:
256…258 °C
Storage Temperature:
Freezer
EINECS:
206-379-0