2,3,5-Triphenyltetrazolium chloride (TTC)
Supplier: Serva
Certificates
About this item
CAS Number:298-96-4
:
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963
Merck Index:13,09815
Boiling Pt:∼360 °C (1013 hPa)
Melting Pt:235 °C
:334.81 g/mol
Storage Temperature:Refrigerator
EINECS:206-071-6
:GHS07