Order Entry
Germany
ContactUsLinkComponent
2,3,5-Triphenyltetrazolium chloride (TTC)
2,3,5-Triphenyltetrazolium chloride (TTC)
Catalog # A6246.0025
Supplier:  PanReac AppliChem
CAS Number:  
2,3,5-Triphenyltetrazolium chloride (TTC)
Catalog # A6246.0025
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:13,09815
Boiling Pt:∼360 °C (1013 hPa)
Melting Pt:235 °C
 :334.81 g/mol
Storage Temperature:Refrigerator
EINECS:206-071-6
 :GHS07
CAS Number:298-96-4
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963