ContactUsLinkComponent
DNMT colorimetric drug discovery tests
DNMT colorimetric drug discovery tests
Catalog # ENZOENZ450170001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
DNMT colorimetric drug discovery tests
Catalog # ENZOENZ450170001
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    DNMT colorimetric drug discovery tests
  • Seq:
    02
Specifications
About this item
DNMT colorimetric drug discovery kit is a simple method to detect DNA methylation activity or inhibition.
  • Easy colorimetric assay without radioactivity or chromatography
  • Measure as little as 0,5 μg of nuclear extract or 0,5 ng of purified enzymes
  • Optimised for high specificity to 5-mC without cross-reactivity to unmethylated cytosine
  • Reproducible results in less than four hours
Suitable for the quantitative determination of DNMT activity/inhibition in nuclear extracts or purified enzymes.
Delivery: Includes wash buffer, DNMT assay buffer, adomet, DNMT enzyme control, capture Ab, detection Ab, enhancer solution, developer solution, stop solution, eight-well assay strips with frame, adhesive covering film.