Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1.000 mL
Catalog # SERA42557.01
Marketsource
Supplier: Serva
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # SERA42557.01
Supplier: Serva
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)



