

Silver nitrate 5% in aqueous solution analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
EINECS:231-853-9
:GHS05
:GHS09
Storage Temperature:Ambient
Frequently Bought Together