0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Ammonium thiocyanate
Ammonium thiocyanate 6% in aqueous solution, extra pure
Ammonium thiocyanate 6% in aqueous solution, extra pure
Ammonium thiocyanate 6% in aqueous solution, extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Ammonium thiocyanate 6% in aqueous solution, extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
1762-95-4
:
Formula:
NH₄SCN
MDL Number:
MFCD00011428
:
76.12 g/mol
Storage Temperature:
Ambient
EINECS:
217-175-6