Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium thiocyanate
Ammonium thiocyanate 12% in aqueous solution analytical reagent
Ammonium thiocyanate 12% in aqueous solution analytical reagent
Ammonium thiocyanate 12% in aqueous solution analytical reagent
Supplier:
Bernd Kraft
Certificates
Expand image
Ammonium thiocyanate 12% in aqueous solution analytical reagent
Supplier:
Bernd Kraft
About this item
CAS Number:
1762-95-4
:
Formula:
NH₄SCN
MDL Number:
MFCD00011428
:
76.12 g/mol
Storage Temperature:
Ambient
EINECS:
217-175-6