Order Entry
Germany
ContactUsLinkComponent
Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Corrosive
Irritant
Environment
Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric, NH₃):
    25.0 - 30.0 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Density (20/20°C):
    0.892 - 0.910
  • Carbonate (as CO₂):
    ≤ 0.0060 %
  • Chloride (Cl):
    ≤ 0.00010 %
  • Sulfate (SO₄):
    ≤ 0.0005 %
  • Pyridine and related substances:
    ≤ 0.0002 %
  • Cu (Copper):
    ≤ 0.00005 %
  • Fe (Iron):
    ≤ 0.000025 %
  • Oxidizable substances:
    passes test
  • Substances reducing potassium permanganate (as O):
    ≤ 0.00075 %
  • Residual solvents (ICH Q3C):
    ...excluded by production process
  • Evaporation residue:
    ≤ 0.0018 %
  • Residue on ignition (as SO₄):
    ≤ 0.002 %
  • Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
    ≤ 0.002 %

Specification Test Results

Related Information