Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 25%, extra pure

Ammonia solution 25%, extra pure

Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 25%, extra pure
Supplier:  Bernd Kraft
Certificates
Ammonia solution 25%, extra pure

4 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 43,80 €
    43,80 €
    1 * 2.500 mL
       43,80 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 24,50 €
    24,50 €
    1 * 1 L
       24,50 €

  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 74,80 €
    74,80 €
    1 * 5 L
       74,80 €

  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 124,00 €
    124,00 €
    1 * 10 L
       124,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.