Ammonia solution 10%, extra pure
Supplier: Bernd Kraft
Jump to:
About this item
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Density:0.92 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
4 Options Available Below
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Density:0.92 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



