Order Entry
Germany
ContactUsLinkComponent
Tetrabutylammonium chloride 50% in aqueous solution
Tetrabutylammonium chloride 50% in aqueous solution
Catalog # MOLE44513467-100ML
Supplier:  Molekula
CAS Number:  
Tetrabutylammonium chloride 50% in aqueous solution
Catalog # MOLE44513467-100ML
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :277.92 g/mol
EINECS:214-195-7
CAS Number:1112-67-0
Formula:C₁₆H₃₆ClN
MDL Number:MFCD00011635