Order Entry
Germany
ContactUsLinkComponent
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
Corrosive
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Corrosive
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.11 g/cm3 (20 °C)
  • pH:
    13.8 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information