
Sulphuric acid ≥90-91% for the determination of fats according to Gerber
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Density:1.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05