Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silver sulphate
Silver sulphate 80 g/l in sulphuric acid
Silver sulphate 80 g/l in sulphuric acid
Silver sulphate 80 g/l in sulphuric acid
KRAF05239.3700
:
Bernd Kraft
:
Silver sulphate 80 g/l in sulphuric acid
KRAF05239.3700
:
Bernd Kraft
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
311.8 g/mol
:
233-653-7
:
1830
:
8,II
:
GHS05
:
10294-26-5
:
Ag₂SO₄
:
MFCD00003407