Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodine
Iodine 0.05 mol/l (0.1 N) in aqueous solution
Iodine 0.05 mol/l (0.1 N) in aqueous solution
Iodine 0.05 mol/l (0.1 N) in aqueous solution
Supplier:
Bernd Kraft
Certificates
Expand image
Iodine 0.05 mol/l (0.1 N) in aqueous solution
Supplier:
Bernd Kraft
About this item
CAS Number:
7553-56-2
:
Formula:
I₂
MDL Number:
MFCD00011355
:
253.81 g/mol
Storage Temperature:
Ambient
EINECS:
231-442-4
UN:
3264
ADR:
8,II