Order Entry
Germany
ContactUsLinkComponent
Calcium chloride 0.025 mol/l (0.05 N) in aqueous solution
Calcium chloride 0.025 mol/l (0.05 N) in aqueous solution
Catalog # BAYR10446232
Supplier:  BAYER VITAL
CAS Number:  
Calcium chloride 0.025 mol/l (0.05 N) in aqueous solution
Catalog # BAYR10446232
Supplier:  BAYER VITAL
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    150 mL
  • Seq:
    2500

Specifications

About this item

 :
 :110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903