


Hydrobromic acid 33% (w/w) in glacial acetic acid, pure
:10035-10-6
:HBr
:MFCD00011323
:80.91 g/mol
:1.42 g/cm³
:39 °C
:Ambient
:233-113-0
:2920
:8,II
:GHS07
:GHS05
:GHS02
- Appearance:Clear yellow to yellow-orange Solution (May darken on storage)
- Water:≤0.5 % (K.F.)
- Titration with NaOH:31.5 to 36.5 % (HBr)
- Particle size:≤1 % (> 250 µm)
- Residue after ignition:≤0.05 %
- pH:5 to 7
- Heavy metals (as Pb):≤10 ppm