

Hydrobromic acid 48% in water, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323
:80.91 g/mol
Melting Pt:–11 °C
Density:1.48 g/cm³
Storage Temperature:Ambient
EINECS:233-113-0
UN:1788
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear light yellow to orange Solution (May darken on storage)
- Titration with NaOH:≥47 %
- Melting point:62 to 66 °C
- GC:≥96.0 %