Order Entry
Germany
ContactUsLinkComponent
Hydrobromic acid 47%, EMSURE® ISO for analysis, Supelco®
Corrosive
Irritant
Hydrobromic acid 47%, EMSURE® ISO for analysis, Supelco®
Catalog # 1.00307.1000
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Hydrobromic acid 47%, EMSURE® ISO for analysis, Supelco®
Catalog # 1.00307.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -11 °C
  • Boiling Point:
    126 °C (1013 hPa)
  • Vapor Pressure:
    10.6 hPa (20 °C)
  • Density:
    1.49 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly acid

Specification Test Results

About this item

 :
 :80.91 g/mol
Density:1.47 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1788
ADR:8,II
 :M-Clarity™ quality level = MQ300
EINECS:233-113-0
 :GHS05
 :GHS07
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323