Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphuric acid
Sulphuric acid 5 mol/l (10 N) in aqueous solution
Sulphuric acid 5 mol/l (10 N) in aqueous solution
Corrosive
Sulphuric acid 5 mol/l (10 N) in aqueous solution
Supplier:
Bernd Kraft
Certificates
Expand image
Corrosive
Sulphuric acid 5 mol/l (10 N) in aqueous solution
Supplier:
Bernd Kraft
About this item
CAS Number:
7664-93-9
:
Formula:
H₂SO₄
MDL Number:
MFCD00064589
:
98.08 g/mol
Storage Temperature:
Ambient
EINECS:
231-639-5
UN:
2796
ADR:
8,II
:
GHS05