

Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®
Supplier: Merck
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,
:36.46 g/mol
:GHS05
:GHS07
Specification Test Results
- Concentration (HCl):5.7 - 6.3 mol/l
- Identity:Passes Test
- Color:≤10 Hazen
- Appearance:Passes Test
- Free Bromine or Free Chlorine:Passes Test
- Free Chlorine (Cl):≤0.0001
- Sulfate (SO₄):≤0.0005%
- Sulfite (SO₃):≤0.001%
- Heavy Metals (as Pb):≤0.0001%
- As (Arsenic):≤0.00015%
- Residual Solvents (ICH Q3C):Excluded by Production Process
- Residue on Ignition (as SO₄):≤0.008%
- Evaporation Residue:≤0.01
- Endotoxins:≤0.25 I.U./ml
Specification Test Results
Frequently Bought Together