

Hydrochloric acid 5 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Catalog # 4.80791.5000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
About this item
:
Density:1.06 g/cm³ (20 °C)
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324