Mercaptoacetic acid (thioglycolic acid) 10% in aqueous solution
Catalog # KRAF15491.3700
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:92.12 g/mol
Storage Temperature:Refrigerator
EINECS:200-677-4
CAS Number:68-11-1
Formula:HSCH₂COOH
MDL Number:MFCD00004876