Acetic acid 60%, TECHNICAL
Catalog # 20127.366
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay:59 - 61 %
Specification Test Results
About this item
:
:60.05 g/mol
Density:1.06
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,II
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152