Pyruvic acid ≥80% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:127-17-3
:
Formula:C₃H₄O₃
MDL Number:MFCD00002585
:88.06 g/mol
:M-Clarity™ quality level = MQ300
Storage Temperature:Refrigerator
EINECS:204-824-3