Hydrochloric acid standardised solution 0.05 N
Glass bottle, 1 L
CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colourless
: Hydrochloric acid solution, Muriatic acid
- :7647-01-0
- :HCl
- :MFCD00011324
- :36.46 g/mol
- :1.004 g/cm³ (25 °C)
- :Ambient
- :231-595-7
- :1789
- :8,III
- Pack type:Glass bottle
- Pk:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



