Order Entry
Germany
ContactUsLinkComponent
Hydrochloric acid 0.02 mol/l (0.02 N)
Hydrochloric acid 0.02 mol/l (0.02 N)
Catalog # MSPP80011986.5000
CAS Number:  
Hydrochloric acid 0.02 mol/l (0.02 N)
Catalog # MSPP80011986.5000
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324