Acetic acid 1 mol/l (1 N), Titripur® Ph. Eur. volumetric solution, Supelco®
Catalog # 1.99061.9025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic can
- Pk:25 L
- Seq:5327
Specifications
About this item
:
:60.05 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-580-7
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152