0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Acetic acid
Acetic acid 1 mol/l (1 N)
Acetic acid 1 mol/l (1 N)
:
Bernd Kraft
Acetic acid 1 mol/l (1 N)
:
Bernd Kraft
:
64-19-7
:
:
H₃CCOOH
:
MFCD00036152
:
60.05 g/mol
:
Ambient
:
200-580-7