Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetic acid
Acetic acid standardised solution 0.1 N
Acetic acid standardised solution 0.1 N
Acetic acid standardised solution 0.1 N
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Acetic acid standardised solution 0.1 N
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
64-19-7
:
Formula:
H₃CCOOH
MDL Number:
MFCD00036152
:
60.05 g/mol
Storage Temperature:
Ambient
EINECS:
200-580-7
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA035567
Videos
Thermo Scientific Acroseal 25 Years
Documentation