(±)-1-Phenoxy-2-propanol tech. 85%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:770-35-4
:
Formula:C₆H₅OCH₂CH(OH)CH₃
MDL Number:MFCD00016861
Boiling Pt:242…244 °C
Melting Pt:11.4 °C
Density:1.060
Flash Pt:120 °C (248 °F)
:152.19 g/mol
Storage Temperature:Ambient
EINECS:212-222-7