Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-2-Methylvaleric acid
(±)-2-Methylvaleric acid, Sigma-Aldrich®
(±)-2-Methylvaleric acid, Sigma-Aldrich®
:
Merck
(±)-2-Methylvaleric acid, Sigma-Aldrich®
:
Merck
:
97-61-0
:
:
C₆H₁₂O₂
:
MFCD00002671
:
195 °C (1013 hPa)
:
–85 °C
:
0.92 g/cm³ (20 °C)
:
87 °C
:
202-594-9
:
3265
:
8,III
:
M-Clarity™ quality level = MQ300
:
116.16 g/mol
:
GHS05