Order Entry
Germany
ContactUsLinkComponent
(±)-2-Butanol
(±)-2-Butanol
Catalog # EHERC10861600
Supplier:  EHRENSTORFER
CAS Number:  
undefined
(±)-2-Butanol
Catalog # EHERC10861600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :74.12 g/mol
Merck Index:13,01540
Boiling Pt:94 °C (1013 hPa)
Melting Pt:–115 °C
Density:0.8534 g/cm³ (20 °C)
Flash Pt:23 °C
Storage Temperature:Ambient
EINECS:201-158-5
UN:1120
ADR:3,III
 :GHS02
 :GHS07
CAS Number:78-92-2
Formula:H₃CCH₂CH(OH)CH₃
MDL Number:MFCD00004569