Order Entry
Germany
ContactUsLinkComponent
(±)-1-Methoxy-2-propanol
Flammable
Irritant
(±)-1-Methoxy-2-propanol
Catalog # EHERC15083000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Flammable
Irritant
(±)-1-Methoxy-2-propanol
Catalog # EHERC15083000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
Boiling Pt:119.6 °C (1013 hPa)
Melting Pt:–97 °C
Density:0.92 g/cm³ (20 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:203-539-1
UN:3092
ADR:3,III
 :90.12 g/mol
 :GHS02
 :GHS07
CAS Number:107-98-2
Formula:CH₃CH(OH)CH₂OCH₃
MDL Number:MFCD00004537