

(±)-1-Methoxy-2-propanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-98-2
:
Formula:CH₃CH(OH)CH₂OCH₃
MDL Number:MFCD00004537
Boiling Pt:119…120 °C
Melting Pt:–100 °C
Density:0.926
Flash Pt:35 °C (95 °F)
Storage Temperature:Ambient
EINECS:203-539-1
ADR:3,III
:90.12 g/mol
UN:3092
:GHS02
:GHS07