Styrene 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-42-5
:
Formula:C₈H₈
MDL Number:MFCD00008612
:104.15 g/mol
Merck Index:13,08944
Boiling Pt:145.2 °C (1013 hPa)
Melting Pt:–31 °C
Density:0.906 g/cm³
Flash Pt:31 °C
Storage Temperature:Refrigerator
EINECS:202-851-5
UN:2055
ADR:3,III
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5460 to 1.5480 (20°C, 589 nm)
- GC:≥99.4 %
- Color scale:≤20 APHA
- Stabilizer:10 to 20 ppm p-tert-Butylcatechol
- Aldehyde:≤0.02 % (as benzaldehyde)
- Polymer:≤10 ppm
- Peroxides:≤0.01 % (as H2O2)