Order Entry
Germany
ContactUsLinkComponent
(±)-1-Aminopropan-2-ol mixture of isomers ≥98% for synthesis, Sigma-Aldrich®
(±)-1-Aminopropan-2-ol mixture of isomers ≥98% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
(±)-1-Aminopropan-2-ol mixture of isomers ≥98% for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Assay isomers (GC, area%):
    ≥ 98.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.959 - 0.961
  • 1-amino-2-propanol (GC, area%):
    ≥ 90.0 % (a/a)
  • Identity (IR):
    passes test

Specification Test Results

Related Information