(±)-1-Aminopropan-2-ol 94% remainder 2-amino-1-propanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-96-6
:
Formula:C₃H₉NO
MDL Number:MFCD00008139
:75.11 g/mol
Boiling Pt:160 °C
Melting Pt:1 °C
Density:0.959
Flash Pt:71 °C (159 °F)
Storage Temperature:Ambient
EINECS:201-162-7
UN:2735
ADR:8,II
:GHS05
:GHS07