Order Entry
Germany
ContactUsLinkComponent
Diethyl ether, anhydrous ≥99% stabilised ACS, Sigma-Aldrich®
Flammable
Irritant
Diethyl ether, anhydrous ≥99% stabilised ACS, Sigma-Aldrich®
Catalog # SIAL673811-6X1L
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether, anhydrous ≥99% stabilised ACS, Sigma-Aldrich®
Catalog # SIAL673811-6X1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    6 L
  • Seq:
    5198

Specifications

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646