Orders LinkContactUsLinkComponent
Diethyl ether stabilised, extra pure
Diethyl ether stabilised, extra pure
Diethyl ether stabilised, extra pure
Diethyl ether stabilised, extra pure
 :  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  •  :60-29-7
  •  :(CH₃CH₂)₂O
  •  :MFCD00011646
  •  :74.12 g/mol
  •  :12,03852
  •  :35 °C (1013 hPa)
  •  :–123 °C
  •  :0.7135 g/cm³ (20 °C)
  •  :–40 °C
  •  :1155
  •  :3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 7 ppm
  •  :Ambient
  •  :200-467-2
  •  :01-2119535785-29
Product Family Options
  • Pack type
    Pk
  • Glass bottle
    500 mL
    1 * 500 mL
    1 * 500 mL - 33,70 €
    33,70 €
    1 * 500 mL
       33,70 €
  • Glass bottle
    2.500 mL
    1 * 2.500 mL
    1 * 2.500 mL - 99,20 €
    99,20 €
    1 * 2.500 mL
       99,20 €
  • Glass bottle
    1 L
    1 * 1 L
    1 * 1 L - 50,50 €
    50,50 €
    1 * 1 L
       50,50 €
  • 5 L
    1 * 5 L
    1 * 5 L - 173,00 €
    173,00 €
    1 * 5 L
       173,00 €
  • 25 L
    1 * 25 L
    1 * 25 L - 553,00 €
    553,00 €
    1 * 25 L
       553,00 €