DL-Sodium lactate, Sigma-Aldrich®
Catalog # SIALL7900-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:112.06 g/mol
Merck Index:13,08709
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-772-0
CAS Number:72-17-3
:312-85-6
Formula:H₃CCH(OH)COONa
MDL Number:MFCD00065400