Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Sodium lactate
DL-Sodium lactate, Sigma-Aldrich®
DL-Sodium lactate, Sigma-Aldrich®
DL-Sodium lactate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
DL-Sodium lactate, Sigma-Aldrich®
Supplier:
Merck
Certificates
About this item
CAS Number:
72-17-3
:
312-85-6
Formula:
H₃CCH(OH)COONa
MDL Number:
MFCD00065400
:
112.06 g/mol
Merck Index:
13,08709
Storage Temperature:
Ambient
EINECS:
200-772-0
Related Information
Documentation
Certificates
Search by Lot Number
Documentation