L(+)-Cysteine hydrochloride
Catalog # MOLE29268471-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:157.62 g/mol
Merck Index:13,02810
Melting Pt:175 °C
Storage Temperature:Ambient
EINECS:200-157-7
CAS Number:52-89-1
Formula:HSCH₂CH(NH₂)COOH·HCl
MDL Number:MFCD00064553