Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Marketsource
Supplier:   Serva
Tween® 20 (Polysorbate)
Supplier:  Serva
Marketsource
Certificates
Tween® 20 (Polysorbate)

2 Options Available Below

Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 51,10 €
    51,10 €
    1 * 500 g
       51,10 €

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 285,00 €
    285,00 €
    1 * 5 kg
       285,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.